Learn More
TMEM192 Rabbit anti-Human, Unconjugated, Polyclonal, Proteintech
Artikelnummer. 16886065 Alle ansehen Proteintech Produkte
Change view
Click to view available options
Menge:
20 μL
150 μL
Unit Size:
150 Mikroliter
20 Mikroliter
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge unitSize
16886065 20 μL 20 Mikroliter
16876065 150 μL 150 Mikroliter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16886065 Supplier Proteintech Supplier No. 282631AP20UL
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Rabbit Polyclonal Antibody
The TMEM192 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human TMEM192. This antibody recognizes human, mouse antigen. The TMEM192 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Antigen TMEM192
Anwendungen Western Blot, Immunohistochemistry, ELISA
Klassifikation Polyclonal
Konzentration 367 μg/mL
Konjugat Unconjugated
Wirtsspezies Rabbit
Menge 20 μL
Primär oder sekundär Primary
Gen-ID (Entrez) 201931
Zielspezies Human, Mouse
Isotype IgG