missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMBIM 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marke: Novus Biologicals NB110-87040
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TIMBIM 1 Polyclonal specifically detects TIMBIM 1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry.
Spezifikation
| TIMBIM 1 | |
| Polyclonal | |
| Unconjugated | |
| 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide | |
| TMBIM 1, TMBIM1, Transmembrane BAX inhibitor motif containing 1, Transmembrane BAX inhibitor motif containing protein 1 | |
| This protein A purified TIMBIM 1 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant protein corresponding to the amino terminus of human TIMBIM 1 protein. (Uniprot: Q969X1) | |
| Protein A purified | |
| RUO | |
| This is specific for human TMBIM1 protein. | |
| Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Dot Blot | |
| 1.2 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000, Immunohistochemistry 10ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml, Immunohistochemistry-Paraffin 1:50 - 1:200, Dot Blot | |
| Q969X1 | |
| Rabbit | |
| 34.61 kDa | |
| 0.1 mg | |
| Primary | |
| Human | |
| Purified |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur