missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombomodulin / CD141 Mouse, Clone: THBD/1591, enQuireBio™
Mouse Monoclonal Antibody
Brand: EnquireBio 7056-MSM1-P1
Additional Details : Gewicht : 0.01000kg
Specifications
Thrombomodulin / CD141 | |
Cell Surface and Cytoplasmic | |
THBD/1591 | |
Unconjugated | |
NP_000352 | |
Mouse | |
100kDa | |
Protein A/G Purified | |
RUO | |
7056 | |
IgG2b κ |
Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
THBD | |
Recombinant human Thrombomodulin protein fragment (aa69-194) | |
THP1 Cells. Placenta, Angiosarcoma, Bladder or Cervical Carcinoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |