missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thrombomodulin / CD141 (Endothelial Cell Marker) Mouse, Clone: rTHBD/1591, enQuireBio™
Mouse Monoclonal Antibody
Marke: EnquireBio 7056-MSM3-P1
Additional Details : Gewicht : 0.01000kg
Spezifikation
Thrombomodulin / CD141 (Endothelial Cell Marker) | |
Cell Surface and Cytoplasmic | |
rTHBD/1591 | |
Unconjugated | |
Mouse | |
∽100kDa | |
Protein A/G purified from Bioreactor concentrate. | |
RUO | |
7056 | |
IgG1 |
ELISA, Flow Cytometry, Immunofluorescence, Western Blot, Immunohistochemistry | |
Monoclonal | |
200μg/mL | |
10mM PBS with 0.05% BSA and 0.05% azide. Also available without BSA and Azide. | |
Recombinant human Thrombomodulin protein fragment (apprx. aa 69-194) | |
THP1 Cells. Placenta, Angiosarcoma, Bladder or Cervical Carcinoma | |
100 μg | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. |