missing translation for 'onlineSavingsMsg'
Learn More
Learn More
THRB Human Protein, Ligand Binding Domain, GST-Tagged
THRB Human Protein, Ligand Binding Domain, GST-Tagged
Marke: Invitrogen PV4764
Additional Details : Gewicht : 0.50000kg
Beschreibung
Thyroid Receptor beta, Ligand Binding Domain (THRB LBD) is a 57.40 kDa recombinant human protein (amino acids 202-461) expressed as a GST-tagged fusion protein in baculovirus-infected insect cells. THRB LBD was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies.Spezifikation
NP_000452.2 | |
Pharma and Biopharma, Target and Lead Identification and Validation, Ligand Binding Assay | |
THRB Human Protein | |
–68 to –85°C | |
1 Each | |
THRB Human Protein | |
THRB Human Protein | |
Hormone receptors |
0.36mg/mL | |
57.4kDa | |
50μg | |
For research use only. Not for use in diagnostic procedures | |
TR-β LBD | |
Amino acids 202-461 | |
Human | |
85% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.