missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Thioglycollate Medium USP

Artikelnummer. 12697218
Change view
Click to view available options
Füllgewicht:
100, bis 101,5 g
100.0 bis 101,5 g
20 bis 20,3 g
500 bis 504 g
9 ± 0.5 ml
Format:
100 ml-Fläschchen
125 ml-Sirupflasche
500 ml DIN-Flasche
Tube
Universalflasche
Packungsgröße:
1 Stück
10 Stück
50 Stück
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Füllgewicht Format unitSize
12697218 100.0 bis 101,5 g 100 ml-Fläschchen -
13295789 9 ± 0.5 ml Tube 50 Stück
12697198 20 bis 20,3 g Universalflasche 1 Stück
13265439 100, bis 101,5 g 125 ml-Sirupflasche 10 Stück
13275489 500 bis 504 g 500 ml DIN-Flasche 10 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 12697218 Lieferant Thermo Scientific™ Lieferanten-Nr. BO0368M

Please to purchase this item. Need a web account? Register with us today!

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Cultivate aerobic and anaerobic organisms in the performance of sterility tests with the convenience of prepared media tubes.

Cultivate aerobic and anaerobic organisms in the performance of sterility tests with Thermo Scientific™ Thioglycollate Medium USP prepared media tubes. The medium is prepared according to the formula specified in the US Pharmacopoeia1 for the performance of sterility tests, and conforms to formulations detailed in the British and European Pharmacopoeia 2,3.

Thioglycollate Medium USP is suitable for the cultivation of both aerobic and anaerobic organisms. No paraffin or special seal is necessary nor is an anaerobic jar required for the cultivation of anaerobes. It is well buffered, so that acid or alkaline inocula produce negligible alteration in the reaction of the medium. The sodium thioglycollate content of the medium will neutralize the bacteriostatic effect of mercurial compounds used as preservatives in solutions for injection, etc. If the solution undergoing test contains a bacteriostatic substance, it is necessary, in order to avoid a false negative result, to establish the bacteriostatic activity of the product by the method described in the US Pharmacopoeia1. Thioglycollate Medium USP is also recommended for the cultivation of Clostridium species.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.

TRUSTED_SUSTAINABILITY

Spezifikation

Farbe Straw/Yellow with or without Red Layer
Inhalt und Lagerung 2°C to 25°C
Format 100 ml-Fläschchen
Herstellungsort United Kingdom
Produkttyp Microbiology Media
Füllgewicht 100.0 bis 101,5 g
Verpackungsart 10 x 100 mL in 100 mL vial
Menge 10/Pk

For In Vitro Diagnostic Use.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.