Learn More
Invitrogen™ Human MMP-14 (Prodomain-catalytic domain-hemopexin domain) His-tag Recombinant Protein
Recombinant Protein
Marke: Invitrogen™ RP77533
Additional Details : CAS : 69-74-9 Gewicht : 0.01000kg
Beschreibung
MMP14 Recombinant Protein, RP-77533, from Invitrogen.
RP-77533 is a purified His-tagged recombinant MT1-MMP/MMP-14 protein (prodomain, catalytic domain, and hemopexin domain) expressed in E.coli. The pro-enzyme consists of MT1-MMP amino acid residues Ser1-Val501 followed by one Thr-residue and six His-residues. RP-77533 may be used as a control with corresponding anti-MMP-14 antibodies. The MW of soluble pro-MT1-MMP is 58kDa. The pro-enzyme can be activated with trace amounts of the MT1-MMP catalytic domain.Spezifikation
P50281 | |
TBS with 5mM CaCl2 and no preservative; pH 7.5 | |
Human MMP14 | |
10μg | |
-80°C, Avoid Freeze/Thaw Cycles | |
MT1-MMP | |
MMP14 | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
Control | |
4323 | |
7.5 | |
E. coli | |
RUO | |
Human MMP14 | |
Human | |
Hu | |
Recombinant | |
Recombinant |
in vitro research only