Learn More
Invitrogen™ Human Aggrecan (Interglobular Domain) Recombinant Protein
Recombinant Protein
Marke: Invitrogen™ RP77527
Additional Details : CAS : 69-74-9 Gewicht : 0.01000kg
Beschreibung
Aggrecan Interglobular Domain (human) Recombinant Protein, RP-77527, from Invitrogen.
The human aggrecan amino acid sequence T331-G458 is expressed in E. coli with a C-terminal His-tag. The recombinant protein contains cleavage sites for aggrecanases (E373-A374 in aggrecan) and matrix metalloproteinases (N341-F342 in aggrecan). RP-77527 is a purified recombinant Aggrecan Interglobular Domain protein, amino acid sequence T331-G458 expressed in E. coli with a C-terminal His-tag. The recombinant protein contains cleavage sites for aggrecanases (E373-A374 in aggrecan) and matrix metalloproteinases (N341-F342 in aggrecan). RP-77527 may be used as a control with corresponding anti-Aggrecan Interglobular Domain antibodies.Compliance
in vitro research only.
Spezifikation
P16112 | |
Control | |
176 | |
7.5 | |
E. coli | |
RUO | |
ACAN | |
Unconjugated | |
E. coli | |
Liquid | |
purified |
0.5mg/mL | |
TBS with 5mM CaCl2 and no preservative; pH 7.5 | |
Human Aggrecan (Interglobular Domain) | |
100μg | |
-80°C, Avoid Freeze/Thaw Cycles | |
Human Aggrecan (Interglobular Domain) | |
Human | |
Hu | |
Recombinant | |
Recombinant |