Learn More
Tocris Bioscience™ Thalidomide 4'-ether-PEG3-amine
Click to view available options
Menge:
25 mg
Packungsgröße:
25 Milligramm
Beschreibung
Thalidomide 4'-ether-PEG3-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a short PEG linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTAC R&D.
This product has been recently renamed. The previous name for this product was Thalidomide - linker 11
Please contact us for SD files of our available Degrader Building Blocks.
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Spezifikation
Spezifikation
| Chemischer Name oder Material | 4-[2-[2-[2-(2-Aminoethoxy)ethoxy]ethoxy]ethoxy]-2-(2,6-dioxo-3-piperidinyl)-1 H-isoindole-1,3(2 H)-dione hydrochloride |
| CAS | 2387510-82-7 |
| Synonym | Thalidomide - linker 11 |
| Menge | 25 mg |
| Ziel | Degrader Building Blocks |
| Summenformel | C21N27N3O8.HCl |
| Reinheit | 95% |