Learn More
Tocris Bioscience™ Thalidomide 4'-ether-alkylC6-amine
Click to view available options
Menge:
25 mg
Packungsgröße:
25 Milligramm
Beschreibung
Thalidomide 4'-ether-alkylC6-amine is a functionalized cereblon ligand for PROTAC® research and development; incorporates an E3 ligase ligand plus a C6 alkyl linker ready for conjugation to a target protein ligand. Part of a range of functionalized tool molecules for PROTACs R&D.
This product has been recently renamed. The previous name for this product was Thalidomide - linker 4
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Spezifikation
Spezifikation
| Chemischer Name oder Material | 4-[(6-Aminohexyl)oxy]-2-(2,6-dioxo-3-piperidinyl)-1H-isoindole-1,3(2H)-dione hydrochloride |
| CAS | 2245697-88-3 |
| Synonym | Thalidomide - linker 4 |
| Menge | 25 mg |
| Summenformel | C19H23N3O5.HCl |
| Formelmasse | 409.86 |
| Reinheit | 95% |