missing translation for 'onlineSavingsMsg'
Learn More

TFPI Rabbit anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 20865-1-AP-20UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16850445

  • 133.80 CHF / 20 Mikroliter
Verfügbar ab: 17-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Tissue factor pathway inhibitor (TFPI), also known as lipoprotein-associated coagulation inhibitor, is a protease inhibitor that regulates the tissue factor (TF)-dependent pathway of blood coagulation. TFPI is glycosylated and predomitly found in the vascular endothelium and plasma in both free forms and complexed with plasma lipoproteins. The coagulation process initiates with the formation of a factor VIIa-TF complex, which proteolytically activates additional proteases (factors IX and X) and ultimately leads to the formation of a fibrin clot. TFPI inhibits the activated factor X and VIIa-TF proteases in an autoregulatory loop.
Spezifikation

Spezifikation

TFPI
Polyclonal
Unconjugated
TFPI
EPI, Extrinsic pathway inhibitor, LACI, TFI, TFPI, TFPI1
Rabbit
Antigen Affinity Chromatography
RUO
7035
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P10646
TFPI
TFPI Fusion Protein Ag14913
20 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote