Learn More
Tetracycline, Thermo Scientific™
Tetracycline, Deschlorobiomycin, CAS # 60-54-8, is a broad-spectrum napthacene antibiotic that inhibits bacterial growth.
Marke:  Thermo Scientific Alfa Aesar J61714.06
Artikelnummer. 15414259
  • ONLINE EXCLUSIVE PRICE
    41.60 CHF
    40.47 CHF / 5 Gramm
    Sparen 1.13 CHF
    2% Rabatt
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
General Description
• Tetracycline is a semisynthetic compound derived from chlortetracycline, an isolate from Streptomyces aureofaciens
• Tetracycline is a broad-spectrum bacteriostatic that inhibits polypeptide formation by binding the 30S ribosomal subunit and preventing aminoacyl tRNA-mRNA complex formation
• It may have other bactericidal properties such as limited binding to the 50S subunit
Application
• Tetracycline has been shown to exhibit antiprotozoal properties
• It can be used to kill both gram-positive and gram-negative bacteria
• It is suitable for use in laboratory applications in resistance gene testing and research on infection
Videos
Sicherheitsdatenblatt (SDS) Dokumentation