missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TET3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Marke: Novus Biologicals NBP2-13427
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TET3 Polyclonal specifically detects TET3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Spezifikation
| TET3 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1-4 ug/ml, Immunohistochemistry-Paraffin 1:50-1:200 | |
| EC 1.14.11.n2, hCG_40738, KIAA0401, methylcytosine dioxygenase TET3, MGC22014, probable methylcytosine dioxygenase TET3, tet oncogene family member 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 200424 | |
| Human | |
| IgG |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| TET3 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: SLKGGLSQQGLKPSLKVEPQNHFSSFKYSGNAVVESYSVLGNCRPSDPYSMNSVYSYHSYYAQPSLTSVNGFHSKYALPSFSYYGFPSSNPVFPSQFLGPGAWGHSGSSGSFEKKPDLH | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur