missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TERT Antibody (2D8), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 9 publications
Marke: Novus Biologicals NB100-297
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
TERT Monoclonal specifically detects TERT in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Knockdown Validated.
Spezifikation
| TERT | |
| Monoclonal | |
| Unconjugated | |
| Ascites with 0.1% Sodium Azide | |
| EST2Telomerase-associated protein 2, hEST2, TCS1EC 2.7.7.49, Telomerase catalytic subunit, telomerase reverse transcriptase, TP2HEST2, TRTEC 2.7.7 | |
| Mouse | |
| 127 kDa | |
| 0.1 mL | |
| Autophagy, Cancer, Cellular Markers, Chromatin Research, DNA Repair, DNA replication Transcription Translation and Splicing, Embryonic Stem Cell Markers, Stem Cell Markers | |
| 7015 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. | |
| IgM |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2D8 | |
| Western Blot 1:500-1:1000, Flow Cytometry 1:50-1:200, Immunohistochemistry 1:50-1:100, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:100, Immunohistochemistry-Frozen 1:50-1:100, Knockdown Validated | |
| O14746 | |
| TERT | |
| Full-length recombinant human Telomerase reverse transcriptase from insect cells. [UniProt# O14746] | |
| Unpurified | |
| RUO | |
| Primary | |
| Human | |
| Ascites |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur