missing translation for 'onlineSavingsMsg'
Learn More

TER-119 Monoclonal Antibody (TER-119), Super Bright™ 780, eBioscience™, Invitrogen™

Rat Monoclonal Antibody

168.00 CHF - 389.00 CHF

Spezifikation

Antigen TER-119
Klon TER-119
Konzentration 0.2 mg/mL
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15964184
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
78-5921-82
100 μg
389.00 CHF
100 Mikrogramm
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 10-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15954184
Unterlagen ansehen Aktionsdetails
Affymetrix eBioscience
78-5921-80
25 μg Dieser Artikel wurde eingestellt und ist leider nicht mehr verfügbar. Sehen Sie sich das Produkt an, um alternative Produktvorschläge zu sehen oder kontaktieren Sie unseren Technischen Support unter 056 618 41 11.
N/A Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Beschreibung

Beschreibung

Description: The RB6-8C5 monoclonal antibody reacts with mouse Ly-6G, a 21-25 kDa protein also known as the myeloid differentiation antigen Gr-1. A GPI-linked protein, Gr-1 is expressed by the myeloid lineage in a developmentally regulated manner in the bone marrow. While monocytes only express Gr-1 transiently during their bone marrow development, the expression of Gr-1 on bone marrow granulocytes as well as on peripheral neutrophils is a good marker for these populations. eBioscience testing indicates that in the bone marrow and lysed whole blood, the antibody clone RB6-8C5 also stains cells that express the highest levels of Ly6c (as defined by staining with antibody clone HK1.4). It is recommended that 1A8-Ly6G (cat. 9668) be used when looking at Ly-6G specific targets. Applications Reported: This RB6-8C5 antibody has been reported for use in flow cytometric analysis. Applications Tested: This RB6-8C5 antibody has been tested by flow cytometric analysis of mouse bone marrow cells. This may be used at less than or equal to 0.25 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Super Bright 780 is a tandem dye that can be excited with the violet laser line (405 nm) and emits at 780 nm.

TER-119 Monoclonal antibody specifically detects TER-119 in Mouse samples. It is validated for Flow Cytometry.
Spezifikation

Spezifikation

TER-119
0.2 mg/mL
Monoclonal
Liquid
RUO
104231
IgG2b κ
Affinity chromatography
Antibody
TER-119
Flow Cytometry
Super Bright 780
Rat
Mouse
Ly76
Primary
4° C, store in dark, DO NOT FREEZE!
Ly76
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications