missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TER-119 Rat, FITC, Clone: TER-119, enQuireBio™
Rat Monoclonal Antibody
Marke: EnquireBio QAB76-F-100ug
Additional Details : Gewicht : 0.01000kg
Spezifikation
TER-119 | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Rat | |
100 μg | |
Primary | |
Store at 2-4°C. |
Flow Cytometry | |
TER-119 | |
FITC | |
Ly76 | |
Protein A/G Purified | |
RUO | |
104231 | |
IgG2b κ |