Learn More
TentaGel™ MB-Br, Thermo Scientific Chemicals
Click to view available options
Menge:
1 g
5 g
Verpackung:
Glasflasche
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Für die Synthese fester Träger
Spezifikation
Chemischer Name oder Material | TentaGel™ MB-Br |
Farbe | Weiß bis gelb |
Physikalische Form | Pulver |
Beladen | 0.4 to 0.6mmol/g |
Verpackung | Glasflasche |
Partikelgröße | 140 to 170 Micron |
Menge | 1 g |