missing translation for 'onlineSavingsMsg'
Learn More

TCTN1 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marke:  Thermo Scientific PA559037

Additional Details : Gewicht : 0.02000kg

Artikelnummer. 15786901

  • 656.00 CHF / 100 Mikroliter
Verfügbar ab: 11-04-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

Immunogen sequence: FSACSVPVVT GDSQFCSQKA VIYSLNFTAN PPQRVFELVD QINPSIFCIH ITNYKPALSF INPEVPDENN FDTLMKTSDG FTLNAESYVS FTTKLDIPTA Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 73%.

TCTN1 Polyclonal antibody specifically detects TCTN1 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot.
Spezifikation

Spezifikation

TCTN1
Polyclonal
Unconjugated
TCTN1
G730031O11Rik; JBTS13; RGD1566266; TCTN1; TECT1; tectonic family member 1; tectonic-1; UNQ9369/PRO34160
Rabbit
Antigen affinity chromatography
RUO
79600
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q2MV58
TCTN1
Recombinant protein corresponding to Human TCTN1
100 μL
Primary
Human
Antibody
IgG
Produktvorschläge

Produktvorschläge

Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Zertifikate
Sonderangebote

Sonderangebote

Research Use Only