missing translation for 'onlineSavingsMsg'
Learn More

TCR β chain Hamster anti-Mouse, PE, Clone: H57-597, BD

Hamster Monoclonal Antibody

149.00 CHF - 255.00 CHF

Spezifikation

Antigen TCR β chain
Klon H57-597
Konzentration 0.2mg/mL
Anwendungen Flow Cytometry
Klassifikation Monoclonal
Mehr Spezifikationen ansehen

Produkte 2
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
Produktcode Marke Menge Preis Menge & Verfügbarkeit  
15880749
Unterlagen ansehen Aktionsdetails
BD Biosciences
561081
25 μg
149.00 CHF
1 Stück
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 13-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
15827058
Unterlagen ansehen Aktionsdetails
BD Biosciences
553172
0.1 mg
255.00 CHF
1 Stück
Alternative Produkte anzeigen
Sparen Sie bis zu 
»
Verfügbar ab: 13-05-2024
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen
 
Beschreibung

Beschreibung

The H57-597 antibody reacts with a common epitope of the β chain of the T-cell Receptor (TCR) complex on αβ TCR-expressing thymocytes and peripheral T lymphocytes and NK1.1+ thymocytes and NK-T cells of all mouse strains tested. It does not react with γδ TCR-bearing T cells. In the fetal and adult thymus, the TCR β -chain may form homodimers or pair with the pre-TCR α -chain on the surface of immature thymocytes before expression of the TCR α -chain. Plate-bound or soluble H57-597 antibody activates αβ TCR-bearing T cells, and plate-bound mAb can induce apoptotic death.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Spezifikation

Spezifikation

TCR β chain
0.2mg/mL
Monoclonal
Hamster
Mouse
TCR affinity-purified from mouse T-cell hybridoma DO-11.10
Affinity Purified
H57-597
Flow Cytometry
PE
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation

Dokumentation

Product Certifications

For Research Use Only.