missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ TBARS Parameter Assay Kit
Click to view available options
Menge:
2 x 96-Well-Platten
Packungsgröße:
1 Set
Beschreibung
The TBARS Parameter Assay Kit quantifies multi-species TBARS/Thiobarbituric Acid Reactive Substances in cell culture supernates (150 ul), cell lysates (150 ul), serum (150 ul), edta plasma (150 ul), heparin plasma (150 ul), urine (150 ul).
Spezifikation
Spezifikation
| Assay-Bereich | 0.00 - 16 uM |
| Assay-Empfindlichkeit: | 0.055 uM |
| Konjugat | Biotin |
| Probentyp | Cell Culture Supernatant, Zelllysate, Plasma (EDTA, Heparin), Serum, Urin |
| Spezifität | TBARS in various samples |
| Zur Verwendung mit (Anwendung) | ELISA |
| Menge | 2 x 96-Well-Platten |
| Probenvolumen | Cell Culture Supernates (150uL),Cell Lysates (150uL),Serum (150uL),EDTA Plasma (150uL),Heparin Plasma (150uL),Urine (150uL) |
| Lagerungsbedingungen | Store the unopened product at 2° to 8°C. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction