missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAZ Mouse anti-Human, Mouse, Rat, Clone: 2A12A10, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66500-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
WW domain-containing transcription regulator protein 1 (WWTR1) is a transcriptional coactivator which acts as a downstream regulatory target in the Hippo signaling pathway that plays a pivotal role in organ size control and tumor supression by restricting proliferation and promoting apoptosis. The core of this pathway is composed of a kinase cascade wherein STK3/MST2 and STK4/MST1, in complex with its regulatory protein SAV1, phosphorylates and activates LATS1/2 in complex with its regulatory protein MOB1. In turn, MOB1 phosphorylates and inactivates YAP1 oncoprotein and WWTR1/TAZ. WWTR1 enhances PAX8 and NKX2-1/TTF1-dependent gene activation. WWTR1 regulates the nuclear accumulation of SMADS and has a key role in coupling them to the transcriptional machinery such as the mediator complex. WWTR1 also regulates embryonic stem-cell self-renewal and promotes cell proliferation and epithelial-mesenchymal transition.Spezifikation
TAZ | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9EPK5, Q9GZV5 | |
Wwtr1 | |
TAZ Fusion Protein Ag13330 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
2A12A10 | |
Unconjugated | |
WWTR1 | |
TAZ, WWTR1 | |
Mouse | |
Protein G | |
RUO | |
25937, 295062, 97064 | |
Antibody | |
IgG1 |