Learn More
TATDN3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marke: Thermo Scientific PA556922
Additional Details : Gewicht : 0.02000kg
Beschreibung
Immunogen sequence: AGVGLVDCHC HLSAPDFDRD LDDVLEKAKK ANVVALVAVA EHSGEFEKIM QLSERYNGFV LPCL Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 28%.
TATDN3 Polyclonal antibody specifically detects TATDN3 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Spezifikation
TATDN3 | |
Polyclonal | |
Unconjugated | |
TATDN3 | |
1500010M24Rik; Putative deoxyribonuclease TATDN3; TatD DNase domain containing 3; TATDN3 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
128387 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q17R31 | |
TATDN3 | |
Recombinant protein corresponding to Human TATDN3 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only