missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ TaqMan™ Genexpressionsassay (FAM)

Artikelnummer. 32226620 Alle ansehen Applied Biosystems Produkte
Ansicht ändern
Click to view available options
Menge:
250 Reactions, Inventoried
2900 Reactions, Made to Order
360 Reactions, Made to Order
75 Reactions, Inventoried
75 Reactions, Made to Order
750 Reactions, Made to Order
L (2900 Reactions), Made to Order
M (750 Reactions), Made to Order
S (360 Reactions), Made to Order
Marker oder Farbstoff:
ABY
Cy5
FAM
JUN
Packungsgröße:
1 Stück
360 Mikroliter
18 product options available for selection
Product selection table with 18 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Marker oder Farbstoff Packungsgröße
32226620 M (750 Reactions), Made to Order Cy5 1 Stück
10794597 250 Reactions, Inventoried FAM 1 Stück
11997011 2900 Reactions, Made to Order FAM 1 Stück
11620099 750 Reactions, Made to Order FAM 1 Stück
10330545 360 Reactions, Made to Order FAM 360 Mikroliter
11967021 75 Reactions, Made to Order FAM 1 Stück
11977021 75 Reactions, Inventoried FAM 1 Stück
32044651 S (360 Reactions), Made to Order FAM 1 Stück
32044894 M (750 Reactions), Made to Order FAM 1 Stück
32045134 L (2900 Reactions), Made to Order FAM 1 Stück
32226451 S (360 Reactions), Made to Order ABY 1 Stück
32226518 M (750 Reactions), Made to Order ABY 1 Stück
32045072 L (2900 Reactions), Made to Order ABY 1 Stück
32226608 S (360 Reactions), Made to Order JUN 1 Stück
32226471 M (750 Reactions), Made to Order JUN 1 Stück
32044719 L (2900 Reactions), Made to Order JUN 1 Stück
32226612 S (360 Reactions), Made to Order Cy5 1 Stück
32044728 L (2900 Reactions), Made to Order Cy5 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 18 options available.
18 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 32226620 Lieferant Applied Biosystems™ Lieferanten-Nr. A80723
 Eingeschränkter Artikel
Verfügbar ab: 19-02-2027
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

Applied Biosystems TaqMan Gene Expression assays are used for quantitative real-time PCR or digital PCR analysis of gene expression and consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (FAM) on the 5' end and a minor groove binder (MGB) and non-fluorescent quencher (NFQ) on the 3' end.

Applied Biosystems TaqMan Gene Expression assays are used for quantitative real-time PCR or digital PCR analysis of gene expression and consist of a pair of unlabeled PCR primers and a TaqMan probe with a dye label (FAM, VIC, ABY, JUN, and Cyanine 5) on the 5' end and a minor groove binder (MGB) and non-fluorescent quencher (NFQ) on the 3' end.

Features of TaqMan Gene Expression assays include:

  • Easy to use—just add cDNA and master mix and run—no melt curves required
  • Specific—TaqMan assays use proprietary MGB-containing probes that can be up to 15 bases shorter than non-MGB probes, improving the specificity of the assay
  • Sensitive—TaqMan assays are ideal for measuring low levels of expression or low-abundance targets
  • Accurate—identify small fold-changes with high accuracy of quantitation
  • Extensive content—Over 1.8 million predesigned assays available for over 25 different species
  • Gold-standard TaqMan qPCR chemistry—TaqMan assays draw on Thermo Fisher Scientific's bioinformatics assay design pipeline to help ensure high specificity and minimal cross-reactivity, even for gene variants with high sequence homology
  • Fast delivery—your assays shipped in as little as 1–2 days (see below)
  • Multiplexing ready—TaqMan assays are now available with 5 different dye labels to support simultaneous detection of up to 5 targets in a single qPCR reaction. FAM, ABY, JUN, and Cyanine 5 assays are available with balanced primer/probe concentrations (Primer-Limited, 350 nM) to support consistent multiplex performance. VIC Primer-Limited assays (150 nM) are available with SKU 4448484 (S), 4448485 (M), 4448486 (L).

Approximate ship time

  • Inventoried: 1–2 days in North America, 3–5 days in Europe*
  • Made to order: 4–6 days in North America, 6–10 days in Europe (FAM, VIC); 4-10 days in North America, 6-14 days in Europe (ABY, JUN, Cyanine 5)

TaqMan Gene Expression assays are the gold standard in qPCR and dPCR gene expression studies, built on more than 20 years of experience. Each assay includes target primers and a sequence-specific probe optimized for the best functional performance. No additional design, optimization, or melt curve analysis is needed. Available in a wide variety of formats and species, new assay designs are constantly added to help meet your research needs. TaqMan assays have been cited in over 40,000 publications and are backed by more than 350 patents.

Pre-formulated assay

  • 2 unlabeled PCR primers (Standard: 900 nM each final 1X concentration; Primer-Limited: 350 nM each final 1X concentration)
  • 1 dye-labeled (FAM, VIC, ABY, JUN or Cyanine 5) TaqMan MGB probe (250 nM final 1X concentration)

All of our predesigned TaqMan Gene Expression assays are covered by the TaqMan Assays Guarantee.**

*Our most popular assays are stocked in North America and Europe for overnight delivery where possible. If an assay is out of stock, it may take an additional 3–5 business days for delivery.

**Subject to terms and conditions. For complete details, go to www.thermofisher.com/taqmanguarantee.

Recommended master mix: TaqMan Fast Advanced Master Mix (Cat. No. 4444556) or TaqPath DuraPlex 1-Step RT-qPCR Master Mix (Cat. No. A58666)

TRUSTED_SUSTAINABILITY

Spezifikation

Konzentration 20X (Primer Limited)
Inhalt und Lagerung •1 tube containing 20X (XS, S, and M sizes) or 60X (L size) mix of pre-formulated assay (1 probe and 2 primers).
•Store at -15°C to -25°C.
Nachweisverfahren Primer-probe
Form Liquid
Format Tube
PCR-Methode qPCR
Reaktionsgeschwindigkeit Fast
Zur Verwendung mit (Geräte) 7500 System, 7500 Fast System, 7900HT System, StepOne, StepOnePlus, ViiA 7 System, QuantStudio 3, QuantStudio 5, QuantStudio 6 Flex, QuantStudio 7 Flex, QuantStudio 6 Pro, QuantStudio 7 Pro, QuantStudio 12k Flex, QuantStudio Absolute Q Digital PCR System
Interne Sondenmodifikation Cyanine 5 (5'), MGB (Minor Groove Binder) (3')
Marker oder Farbstoff Cy5
Produktlinie TaqMan
Produkttyp Gene Expression Assay
Menge M (750 Reactions), Made to Order
Versandbedingung Room Temperature
Spezies C. elegans, Cattle/Bovine, Dog/Canine, Drosophila, Horse/Equine, Swine/Porcine, Chicken, Zebrafish, Horse, Arabidopsis, Rabbit, Porcine, Rat, Human, Monkey, Mouse
Zur Verwendung mit (Anwendung) Gene Expression
Anzahl Reaktionen 750 Reactions, Made to Order
Mehr anzeigen Weniger anzeigen

Nur für Forschungszwecke. Nicht zur Verwendung bei diagnostischen Verfahren.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.