Learn More
Invitrogen™ Talin 1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA582162
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
Immunogen sequence: LLYTAKEAGG NPKQAAHTQE ALEEAVQMMT EAVEDLTTTL NEAASAAGVV GGMVDSITQA INQLDEGPMG EPEGSFVDYQ TTMVRTAKAI AVTVQEMVTK SNTSP Highest antigen sequence indentity to the following orthologs: Rat - 100%, Mouse - 99%.
Talin, a multifunctional constituent of cell-substratum attachment sites, is a high molecular weight protein (225-235 kDa) found in variety of tissues and cell types. It is localized at a subset of adherens junctions, specialized cell-cell and cell-matrix associations that are characterized by the presence of filamentous actin at the cytoplasmic face of the junctional complex. In cultured cells, talin is absent from cell-cell junctions and found predominantly at adhesion plaques and in fibrillar streaks underlying cell surface fibronectin. Talin interacts with at least two other proteins that are localized at adhesion plaques, vinculin and integrin.
Spezifikation
| Talin 1 | |
| Polyclonal | |
| Unconjugated | |
| TLN1 | |
| ILWEQ; KIAA1027; talin 1; talin-1; TLN; TLN1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 7094 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y490 | |
| TLN1 | |
| Recombinant protein corresponding to Human TLN1. Recombinant protein control fragment (Product #RP-88634). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |