Learn More
CELLPATH SYSTEM III MICRO BIOPSY CASSETTE
Click to view available options
Material:
Threaded
Packungsgröße:
2000 Stück
Beschreibung
- The locking mechanism of the cassette ensures maximum tissue security during processing.
- The cassette has a precisely engineered writing surface which optimises the print quality on inkjet and thermal printers, including the Primera, Leica and Sakura Printers.
- Key Benefits:
- Compatible with Primera, Leica and Sakura Printers
- Precisely engineered writing surface
- Increased reagent flow
- Integral fixed frangible lid
- High rigidity and stability
Spezifikation
Spezifikation
| Farbe | White |
| Material | Threaded |
| Produkttyp | Cassette |