Learn More
CELLPATH SYSTEM II+ ROUND CASSETTE
Artikelnummer. p-200113605
Loose, 1000 Stück
Click to view available options
Material:
Hoppered
Loose
Threaded
Packungsgröße:
1000 Stück
2000 Stück
3000 Stück
Artikelnummer. 30226249
Marke: CELLPATH EAM080203A
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Edge
The System II+ Round Cassette incorporates a round 2.4 mm diameter holes facilitates increased reagent flow in a traditional cassette.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Spezifikation
Farbe White
Material Loose
Produkttyp Cassette
Berichtigung von Produktinhalten
Name des Produkts
CELLPATH SYSTEM II+ ROUND CASSETTE >