Learn More
CELLPATH SYSTEM II+ MICRO BIOPSY CASSETTE
Artikelnummer. p-200113604
Click to view available options
Material:
Hoppered
Loose
Threaded
Packungsgröße:
1000 Stück
2000 Stück
3000 Stück
Artikelnummer. 30226246
Marke: CELLPATH EAM070203A
om dit product te kopen Registreer vandaag om een webaccount aan te maken
The System II+ Micro Biopsy Cassette incorporates an inner chamber with 0.38 x 0.38 mm apertures. CellPath's System II+ Cassettes come with a separate white lid that is easily attached via the use of a hinge.
  • Read rates
  • Robust and secure closure ensures specimen security
  • Designed to improve reagent flow
  • Large color range
  • Writing surface suitable for marking with a CellMark pen
Spezifikation
Farbe White
Material Loose
Produkttyp Cassette