Learn More
Cellpath System II Hex™ Tissue Processing Cassettes (Hoppered)
Pre-loaded into hoppers for use with automated and carousel type printers. CellPath System II Hex Tissue Processing Cassettes (Hoppered) feature unique honeycomb design that increases the open surface area compared to traditional round perforations, allowing increased reagent flow for improved processing and a more rigid block when sectioning.
130.58 CHF
Spezifikation
Menge | 1500/Pk. |
---|---|
Material | Acetyl Resin |
Produkttyp | Tissue Processing Cassette in Hopper tube |
Produktcode | Marke | Farbe | Preis | Menge & Verfügbarkeit | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Farbe | Preis | Menge & Verfügbarkeit | ||||||
30226229
|
Cellpath
EAD-0102-04H |
White |
130.58 CHF
1500 Stück |
Lager vorrätig
Loggen Sie sich ein, um den Lagerbestand zu sehen ![]() |
Sie haben bereits ein Konto? Melden Sie sich an um den Preis und die Verfügbarkeit zu sehen. Neu hier? Registrieren Sie sich noch heute bei uns! |
|||||
Beschreibung
- Pre-loaded into hoppers for use with automated and carousel type cassette printers
- Manufactured from high–grade virgin acetyl resin
- Hexagonal perforations arranged in honeycombed structure to better utilize the available surface area
- Unique arrangement increases open area thereby increasing reagent flow through and improving specimen staining and analysis time
- Extra thick side walls increases rigidity and stability for high quality sectioning
- Designed to suit Thermo Printmate and Micro-writer cassette–writing machines
- Available in 15 stock colors which color code different types of workload or applications
Spezifikation
1500/Pk. | |
Acetyl Resin | |
Tissue Processing Cassette in Hopper tube |