missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Syntaxin 1A / Syntaxin 1B Mouse anti-Human, Mouse, Rat, Clone: 2D3B5, Proteintech
Mouse Monoclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | Syntaxin 1A / Syntaxin 1B |
---|---|
Klon | 2D3B5 |
Konzentration | 1 mg/mL |
Anwendungen | Immunohistochemistry (Paraffin), Western Blot |
Klassifikation | Monoclonal |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16838315
|
Proteintech
66437-1-IG-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16828315
|
Proteintech
66437-1-IG-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 08-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
This gene encodes a member of the syntaxin superfamily. Syntaxins are nervous system-specific proteins implicated in the docking of synaptic vesicles with the presynaptic plasma membrane. Syntaxins possess a single C-terminal transmembrane domain, a SNARE domain, and an N-terminal regulatory domain. Syntaxins bind synaptotagmin in a calcium-dependent fashion and interact with voltage dependent calcium and potassium channels via the C-terminal H3 domain. This gene product is a key molecule in ion channel regulation and synaptic exocytosis. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Spezifikation
Syntaxin 1A / Syntaxin 1B | |
1 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
STX1B, STX1B1, STX1B2, syntaxin 1B, Syntaxin 1B1, Syntaxin 1B2 | |
STX1A, Stx1b | |
IgG1 | |
Protein G | |
Antibody |
2D3B5 | |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Mouse, Human, Rat | |
O35526, P32851, P61264, P61265, P61266, Q16623 | |
112755, 116470, 20907, 24923, 56216, 6804 | |
Syntaxin 1A / Syntaxin 1B Fusion Protein Ag10257 | |
Primary | |
-20°C | |
STX1A |