missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ SynaptoRed C2
GREENER_CHOICE

Product Code. 18730308
missing translation for 'orderingAttributeHoverText'
:
1mg
This item is not returnable. View return policy

Product Code. 18730308

missing translation for 'mfr': Tocris Bioscience™ 5118/1

om dit product te kopen Registreer vandaag om een webaccount aan te maken

This item is not returnable. View return policy

Fluorescent dye; stains synaptic vesicles

SynaptoRedTM C2 is a fluorescent dye; stains synaptic vesicles. Becomes fluorescent when incorporated into plasma membrane, used to follow synaptic activity at the synapse or neuromuscular junction. Excitation and emission spectra are ∽ 515 and 640 nm respectively.
TRUSTED_SUSTAINABILITY

Specifications

Menge 1 mg
Produkttyp SynaptoRed™ C2
Reinheit >95%
Molekulargewicht 607.51
Hemmstoffe SynaptoRed™ C2

Nur für Forschungszwecke

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.