missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Synaptopodin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
133.80 CHF - 381.34 CHF
Spezifikation
Antigen | Synaptopodin |
---|---|
Konzentration | 0.23 mg/mL |
Anwendungen | Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry |
Klassifikation | Polyclonal |
Konjugat | Unconjugated |
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
16850665
|
Proteintech
21064-1-AP-20UL |
20 μL |
133.80 CHF
20 Mikroliter |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
16840665
|
Proteintech
21064-1-AP-150UL |
150 μL |
381.34 CHF
150 Mikroliter |
Verfügbar ab: 13-05-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
SYNPO (Synaptopodin) is an actin-associated protein in telencephalic dendrites and renal podocytes. SYNPO is tightly associated with the dendritic spine apparatus, and mice lacking the SYNPO gene lack the apparatus. These SYNPO-null mice also demonstrate an impaired ability to express long-term potentiation as well as deficits in spatial memory tasks, indicating that SYNPO is involved in the regulation of synaptic plasticity. Recent studies suggest that SYNPO is linked to neuronal calcium stores and plays a role in the calcium store-associated ability of neurons to undergo long-term plasticity.Spezifikation
Synaptopodin | |
Immunohistochemistry (Paraffin), Immunofluorescence, Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
Mouse, Human, Rat | |
Q8CC35, Q8N3V7, Q9Z327 | |
104027, 11346, 60324 | |
Synaptopodin Fusion Protein Ag15089 | |
Primary | |
-20°C | |
SYNPO |
0.23 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA1029, synaptopodin, SYNPO | |
Synpo | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |