missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SYK Mouse anti-Human, Clone: 4C4A12, Proteintech
Mouse Monoclonal Antibody
Marke: Proteintech 66721-1-IG-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene encodes a member of the family of non-receptor type Tyr protein kinases. This protein is widely expressed in hematopoietic cells and is involved in coupling activated immunoreceptors to downstream signaling events that mediate diverse cellular responses, including proliferation, differentiation, and phagocytosis. It is thought to be a modulator of epithelial cell growth and a potential tumor suppressor in human breast carcinomas. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.Spezifikation
SYK | |
Monoclonal | |
1 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
P43405 | |
SYK | |
SYK Fusion Protein Ag17654 | |
150 μL | |
Primary | |
Human | |
Liquid |
Western Blot | |
4C4A12 | |
Unconjugated | |
SYK | |
DKFZp313N1010, FLJ25043, FLJ37489, spleen tyrosine kinase, SYK, Tyrosine protein kinase SYK | |
Mouse | |
Protein A | |
RUO | |
6850 | |
Antibody | |
IgG2b |