Learn More
SUV39H1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke:  Proteintech 10574-1-AP-20UL
 Lernen Sie weitere Varianten dieses
Artikelnummer. 16837043
  • 136.48 CHF / 20 Mikroliter
om dit product te kopen Registreer vandaag om een webaccount aan te maken
Beschreibung
Beschreibung
This gene encodes an evolutionarily-conserved protein containing an N-terminal chromodomain and a C-terminal SET domain. The encoded protein is a histone methyltransferase that trimethylates lysine 9 of histone H3, which results in transcriptional gene silencing. Loss of function of this gene disrupts heterochromatin formation and may cause chromosome instability. Alternative splicing results in multiple transcript variants.
Spezifikation
Spezifikation
SUV39H1
Polyclonal
Unconjugated
SUV39H1
H3 K9 HMTase 1, KMT1A, Lysine N methyltransferase 1A, MG44, Su(var)3 9 homolog 1, SUV39H, SUV39H1
Rabbit
Antigen Affinity Chromatography
RUO
20937, 302553, 6839
-20°C
Liquid
Western Blot, Immunoprecipitation
0.26 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43463, O54864
SUV39H1
SUV39H1 Fusion Protein Ag0854
20 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Produktvorschläge
Produktvorschläge
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Dokumentation
Zertifikate
Aktionen
Aktionen