Learn More
Cytiva Superdex™ 200 3.2/300 Prepacked Precision Column
Sensitive and high-resolution gel filtration separations for biomolecules in micropreparative scale
Marke: Cytiva 29036232
Additional Details : Gewicht : 0.27400kg
Beschreibung
- Prepacked with Superdex 200 media
- For biomolecules with molecular weights between 10,000 and 6,00,000
- Spherical composite of cross-linked agarose and dextran
- Supplied with two fingertight 1/16 in. male connectors for connection
- Used for polishing step in purfication process
- With new hardware design for direct connection to ÄAKTATM or other systems
Spezifikation
0.01 to 0.10 mL/min. | |
13 μm | |
Borosilicate Glass | |
Superdex Pre-packed Column |