missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SULT1A3/SULT1A4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA5116182
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SULT1A3/SULT1A4 Polyclonal Antibody for Western Blot, IHC (P)
Found in a segmental duplication on p arm of chromosome 16 giving rise to two identical copies of this gene sharing exons with SLX1A and SLX1B. The ORFs of SULT1A3 and SULT1A4 differ with only a single nucleotide change that does not alter the encoded amino acid. It is not possible to determine whether any individual polymorphism is present within SULT1A3 or SULT1A4 (PubMed:15358107).
Spezifikation
| SULT1A3/SULT1A4 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P0DMM9, P0DMN0 | |
| SULT1A3, SULT1A4 | |
| KLH conjugated synthetic peptide between 71-99 amino acids from the N-terminal region of human SULT1A3/SULT1A4. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| SULT1A3 | |
| aryl sulfotransferase 1A3/1A4; Catecholamine-sulfating phenol sulfotransferase; dopamine-specific sulfotransferase; HAST; HAST3; Monoamine-sulfating phenol sulfotransferase; monoamine-sulfating phenosulfotransferase; M-PST; phenol sulfotransferase 1A5; placental estrogen sulfotransferase; SLX1B-SULT1A4; ST1A3; ST1A3/ST1A4; ST1A4; ST1A5; STM; sulfokinase; sulfotransferase 1A3; Sulfotransferase 1A3/1A4; Sulfotransferase 1A4; sulfotransferase family 1A member 3; sulfotransferase family 1A member 4; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 3; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 4; sulfotransferase, monoamine-preferring; SULT1A3; SULT1A4; thermolabile (monoamine, M form) phenol sulfotransferase; thermolabile phenol sulfotransferase; TL-PST | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 445329, 6818 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur