missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SULT1A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Marke: Invitrogen™ PA597760
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SULT1A3 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
Sulfotransferase enzymes catalyze the sulfate conjugation of many hormones, neurotransmitters, drugs, and xenobiotic compounds. These cytosolic enzymes are different in their tissue distributions and substrate specificities. The gene structure (number and length of exons) is similar among family members. This gene encodes a phenol sulfotransferase with thermolabile enzyme activity. Four sulfotransferase genes are located on the p arm of chromosome 16; this gene and SULT1A4 arose from a segmental duplication. This gene is the most centromeric of the four sulfotransferase genes. Exons of this gene overlap with exons of a gene that encodes a protein containing GIY-YIG domains (GIYD1). Multiple alternatively spliced variants that encode the same protein have been described.
Spezifikation
| SULT1A3 | |
| Polyclonal | |
| Unconjugated | |
| SULT1A3 | |
| aryl sulfotransferase 1A3/1A4; Catecholamine-sulfating phenol sulfotransferase; CSPS; dopamine-specific sulfotransferase; HAST; HAST3; Monoamine-sulfating phenol sulfotransferase; monoamine-sulfating phenosulfotransferase; M-PST; phenol sulfotransferase 1A5; placental estrogen sulfotransferase; ST1A3; ST1A3/ST1A4; ST1A4; ST1A5; STM; sulfokinase; sulfotransferase 1A3; Sulfotransferase 1A3/1A4; Sulfotransferase 1A4; sulfotransferase family 1A member 3; sulfotransferase family, cytosolic, 1A, phenol-preferring, member 3; Sulfotransferase, monoamine-preferring; SULT1A3; SULT1A4; thermolabile (monoamine, M form) phenol sulfotransferase; Thermolabile phenol sulfotransferase; TL-PST | |
| Rabbit | |
| Protein G | |
| RUO | |
| 6818 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| P0DMM9 | |
| SULT1A3 | |
| Recombinant Human Sulfotransferase 1A3 protein (1-295AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur