Learn More
Sulfanilic Acid Standard, For Elementary Analysis, MilliporeSigma™
Artikelnummer. 15853930
Ansicht ändern
Click to view available options
Menge:
5 g
Packungsgröße:
5 Gramm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Menge Packungsgröße
15853930 5 g 5 Gramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Artikelnummer. 15853930 Lieferant Supelco Lieferanten-Nr. 1006840005
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen
missing translation for 'validMainframeMsg'
Spezifikation
Chemischer Name oder Material Sulfanilic Acid Standard
CAS 121-57-3
Verpackung Glass bottle
Summenformel C6H7NO3S
Menge 5 g
Synonym sulfanilic acid, sulphanilic acid, p-aminobenzenesulfonic acid, sulfanilsaeure, aniline-4-sulfonic acid, benzenesulfonic acid, 4-amino, aniline-p-sulfonic acid, aniline-p-sulphonic acid, p-aminophenylsulfonic acid, sulfanilicacid
InChI-Schlüssel HVBSAKJJOYLTQU-UHFFFAOYSA-N
SMILES C1=CC(=CC=C1N)S(=O)(=O)O
IUPAC-Name 4-aminobenzenesulfonic acid
Molekulargewicht (g/mol) 173.186
PubChem CID 8479
ChEBI CHEBI:27500
Formelmasse 173.18. (molar mass)
Physikalische Form Solid