missing translation for 'onlineSavingsMsg'
Learn More

Sulfamonomethoxin, TRC

Artikelnummer. 17984805
Change view
Click to view available options
Menge:
10 mg
100 mg
Unit Size:
10 Milligramm
100 Milligramm
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Menge Unit Size
17984805 100 mg 100 Milligramm
17903555 10 mg 10 Milligramm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17984805 Supplier Toronto Research Chemicals Supplier No. TRCS699145100MG
 Eingeschränkter Artikel
Verfügbar ab: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hochreine organische Moleküle und analytische Standards, strategisch weltweit bereitgestellt, um Innovation und kommerziellen Erfolg zu ermöglichen.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1220-83-3
Summenformel C11 H12 N4 O3 S
Molekulargewicht (g/mol) 280.3
Synonym 4-Amino-N-(6-methoxy-4-pyrimidinyl)-benzenesulfonamide, Sulfanilamide, N1-(6-methoxy-4-pyrimidinyl)- (6CI,8CI), 4-Methoxy-6-sulfanilamidopyrimidine, 4-Sulfanilamido-6-methoxypyrimidine, 6-Methoxy-4-sulfanilamidopyrimidine, 6-Sulfanilamido-4-methoxypyrimidine, DJ 1550, DS 36, Daimeton, Duphadin, ICI 32525, N1-(6-Methoxy-4-pyrimidinyl)sulfanilamide, Ro 4-3476, Sulfamonomethoxin, Sulfamonomethoxine
IUPAC-Name 4-Amino-N-(6-Methoxypyrimidin-4-yl)Benzolsulfonamid
SMILES COc1cc(NS(=O)(=O)c2ccc(N)cc2)ncn1

Specifications

Chemischer Name oder Material Sulfamonomethoxine
Hinweise zur Reinheitsqualität HPLC
Menge 100 mg
InChI-Formel InChI=1S/C11H12N4O3S/c1-18-11-6-10(13-7-14-11)15-19(16,17)9-4-2-8(12)3-5-9/h2-7H,12H2,1H3,(H,13,14,15)
Formelmasse 280.063
Reinheit (%) >95%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.