missing translation for 'onlineSavingsMsg'
Learn More

STXBP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Marke:  Proteintech 11459-1-AP-150UL

Additional Details : Gewicht : 0.00010kg

 Lernen Sie weitere Varianten dieses

Artikelnummer. 16838553

  • 351.68 CHF / 150 Mikroliter

Verfügbar ab: 28-06-2022
Sie sich ein, um den Lagerbestand zu sehen
Zum Warenkorb hinzufügen

Beschreibung

Beschreibung

This gene encodes a syntaxin-binding protein. The encoded protein appears to play a role in release of neurotransmitters via regulation of syntaxin, a transmembrane attachment protein receptor. Mutations in this gene have been associated with infantile epileptic encephalopathy-4. Alternatively spliced transcript variants have been described.
Spezifikation

Spezifikation

STXBP1
Polyclonal
Unconjugated
STXBP1
EIEE4, MUNC18 1, N Sec1, p67, Protein unc 18 homolog 1, Protein unc 18 homolog A, RBSEC1, STXBP1, syntaxin binding protein 1, Unc 18A, UNC18, Unc18 1, UNC18A
Rabbit
Antigen Affinity Chromatography
RUO
20910, 25558, 6812
Antibody
IgG
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.46 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O08599, P61764, P61765
Stxbp1
STXBP1 Fusion Protein Ag1978
150 μL
Primary
Human, Mouse, Rat
Liquid
Videos
Sicherheitsdatenblatt (SDS)
Dokumentation
Zertifikate
Sonderangebote

Sonderangebote