missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STK33 Recombinant Human Protein
STK33 Recombinant Human Protein
Marke: Invitrogen PV4343
Additional Details : Gewicht : 0.00945kg
Beschreibung
STK33 is a serine⁄threonine kinase involved in signal transduction and cellular communication.Spezifikation
NP_112168 | |
Pharma and Biopharma, Protein Assays and Analysis, Target and Lead Identification and Validation, Kinase Assay | |
61.9kDa | |
10μg | |
For research use only. Not for use in diagnostic procedures | |
STK33 | |
STK33 was pre-diluted in enzyme dilution buffer and assayed in 25mM HEPES (pH 7.5), 10mM MgCl2, 0.5mM EGTA, 0.5mM Na3VO4, 5mM β-glycerophosphate, 2.5mM DTT, 0.01% Triton™ X-100, 100μM ATP, 500μg/mL myelin basic protein (MBP) and trace [32P]-γ-ATP for 10 minutes at 30°C | |
STK33 Recombinant Human Protein | |
Human | |
Baculovirus | |
Serine/threonine kinase |
0.42mg/mL | |
65975 | |
STK33 Recombinant Human Protein | |
–80°C | |
1 Each | |
STK33 Recombinant Human Protein | |
CAMK Unique | |
12.5nmoles of phosphate transferred to myelin basic protein (MBP) per minute per mg of total protein at 30°C. Activity determined at a final protein concentration of 8.33μg/mL | |
His Tag | |
Kinase and inhibitors | |
70% by SDS-PAGE |
For Research Use Only. Not for use in diagnostic procedures.