Learn More
STF 118804, Tocris Bioscience™
NAMPT Inhibitors
Marke: Tocris Bioscience 5207/25
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
STF 118804 is a NAMPT inhibitor (nicotinamide phosphoribosyltransferase, visfatin, PBEF1); inhibits NAD+ synthesis from nicotinamide. Potently inhibits the viability of multiple B-cell acute lymphoplastic leukemia (B-ALL) cell lines (IC50 < 10 nM); induces apoptosis in MF-411 cells. Induces regression of ALL xenografts and eliminates leukemia stems cells from bone marrow in mice.Spezifikation
| 4-[5-Methyl-4-[[(4-methylphenyl)sulfonyl]methyl]-2-oxazolyl]-N-(3-pyridinylmethyl)benzamide | |
| 894187-61-2 | |
| >98% | |
| 25 mg |