missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Stealth RNAi™ siRNA Cyclophilin B Control (human)
Stealth RNAi™ siRNA Cyclophilin B Control (human)
Marke: Invitrogen™ 12935142
Additional Details : Gewicht : 0.50000kg
Beschreibung
Stealth RNAi™ siRNA Positive Housekeeping Control Duplexes (human) are ideal for use in RNA interference (RNAi) experiments as a control for assessing knockdown and optimizing RNAi experiments. These controls are: –Designed to efficiently knock-down the intended target –Have been bench tested and are supplied in a ready-to-use formatSpezifikation
-20°C | |
RNAi, transfection | |
Unconjugated | |
250 μL |
Stealth RNAi™ siRNA Cyclophilin B Control, 250μL, Unconjugated Dye, siRNA RNAi Type, Gene Knockdown Positive Control Type, Used for RNAi, Transfection, Cyclophilin B Target Gene, –20°C Storage Temperature, For Research Use Only | |
Control duplex, 1X RNA Annealing/Dilution Buffer | |
Stealth RNAi™ siRNA Cyclophilin B Control (human) | |
Cyclophilin B |
For Research Use Only. Not for use in diagnostic procedures.