missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Stains-All, 96 %
Click to view available options
Menge:
1 g
5g
Beschreibung
General protein and nucleic acid stain, which stains different colors on bands of RNA (bluish purple), DNA (blue) and proteins (red)
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Formelmasse | 559,58g/mol |
Beilstein | 3865213 |
Produkttyp | Färbemittel |
Menge | 1 g |
Assay | 0,96 |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'