Learn More
Labconco™ Stainless Steel Work Surface for XPert™ and RXPert™ Units
Alle ansehen Labconco Corporation ProdukteBeschreibung
- Type 304 Stainless Steel work surfaces have a 1 in. deep front spill trough and fit the dimensions of XPert Systems and Stations.
- 4 ft. width
Spezifikation
Spezifikation
| Produkttyp | Work Surface |
| Farbe | Silver |
| Anzahl Türen | None |
| Anzahl Schubfächer | None |
| Material | Stainless Steel (304) |