missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arex Professional Equipment™ Stainless Steel Table with Frame on Three Sides, 70 x 120 x 85 mm
Constructed completely of stainless steel with level adjustable legs.
Marke: Arex Professional Equipment™ CTT712S
Additional Details : Gewicht : 32.00000kg
Beschreibung
- Stainless steel construction throughout.
- Adjustable legs.
- Horizontal support bars on three sides.
Spezifikation
70 x 120 x 85 mm | |
70 mm | |
Stainless Steel |
85 mm | |
120 mm | |
Stainless Steel |