missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arex Professional Equipment™ Stainless Steel Table with Bottom Shelf, 60 x 200 x 85 mm
Constructed completely of stainless steel with level adjustable legs.
Marke: Arex Professional Equipment™ CTR620S
Additional Details : Gewicht : 45.00000kg
Beschreibung
- Stainless steel construction throughout.
- Adjustable legs.
- Convenient bottom shelf.
Spezifikation
Bottom Shelf | |
85 mm | |
200 mm | |
Stainless Steel |
60 x 200 x 85 mm | |
60 mm | |
Stainless Steel | |
Stainless Steel |