Learn More
Edelstahlpulver, -20 +35 Mesh, Typ 316-L, Thermo Scientific Chemicals
90.22 CHF - 247.53 CHF
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Menge | Preis | Menge & Verfügbarkeit | |||||
11355777
|
Thermo Scientific Alfa Aesar
043460.36 |
500 g |
90.22 CHF
500 Gramm |
Lager vorrätigLoggen Sie sich ein, um den Lagerbestand zu sehen
|
|||||
11345777
|
Thermo Scientific Alfa Aesar
043460.A3 |
2 kg |
247.53 CHF
2 Kilogramm |
Verfügbar ab: 15-07-2024
Loggen Sie sich ein, um den Lagerbestand zu sehen |
|||||
Beschreibung
Stainless Steel gauze, Type 316 is used to woven wire cloth which has extensive use in chemical processing when better corrosion resistance is required than the regular chromium-nickel types. It is also used in medical, pharmaceutical, laboratory, semiconductor, food processing, marine, waste water and water purification equipment.