Learn More
Arex Professional Equipment™ Stainless Steel Bench
Artikelnummer. 11864310
Encompass
Ansicht ändern
Click to view available options
Abmessungen (L x B x H):
1600 x 600 x 850 mm
1600 x 700 x 850 mm
1800 x 600 x 850 mm
2000 x 700 x 850 mm
Packungsgröße:
1 Stück
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Abmessungen (L x B x H) unitSize
11864310 1600 x 600 x 850 mm 1 Stück
11874310 1800 x 600 x 850 mm 1 Stück
11870281 1600 x 700 x 850 mm 1 Stück
11880281 2000 x 700 x 850 mm 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Artikelnummer. 11864310 Lieferant Arex Professional Equipment™ Lieferanten-Nr. CTR616S