Learn More
Aalborg™ Stainless Steel XFM Air Digital Mass Flowmeter
Artikelnummer. 13557330
Click to view available options
Typ:
Massendurchflussmesser
Packungsgröße:
1 Stück
Artikelnummer. 13557330
Marke: Aalborg™ XFM17SVAL6A2/3AB00SS
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Use a local four button teypad and LCD to access all features of the flow meter. Aalborg™ Stainless Steel XFM Air Digital Mass Flowmeter supports automatic sensor zero offset adjustments.
  • Supports up to 23 Engineering Units (including User Defined).
  • Stores calibration data for up to 10 gases.
  • Programmable Totalizer indicates total gas quantity.
  • High and low gas flow Alarm limits with preset delay interval.
  • Two sets of user-programmable electromechanical SPDT relays with latch option.
  • User-selectable analog 0-5 Vdc or 4-20mA outputs.
  • Internal Conversion factors for up to 32 gases.
  • Digital Interface (RS-232 / RS-485, Profibus DP available).
  • Multi-Drop Capability of up to 256 units (RS-485 option).
  • Optional Profibus DP interface with I&M functionality.
  • Automatic sensor zero offset adjustment (via digital interface or local push button).
  • Self-Diagnostic Tests.
  • Local 2 x 16 characters LCD display with adjustable back light (optional).
Spezifikation
Typ Massendurchflussmesser
Genauigkeit ± 1 %
Luftstromrate 0 to 5 mL/min.
Gastyp Air