missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ST5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marke: Proteintech 12893-1-AP-150UL
Additional Details : Gewicht : 0.00010kg
Beschreibung
This gene was identified by its ability to suppress the tumorigenicity of Hela cells in nude mice. The protein encoded by this gene contains a C-terminal region that shares similarity with the Rab 3 family of small GTP binding proteins. This protein preferentially binds to the SH3 domain of c-Abl kinase, and acts as a regulator of MAPK1/ERK2 kinase, which may contribute to its ability to reduce the tumorigenic phenotype in cells. Three alternatively spliced transcript variants of this gene encoding distinct isoforms are identified.Spezifikation
ST5 | |
Polyclonal | |
Unconjugated | |
ST5 | |
DENND2B, HeLa tumor suppression 1, HTS1, p126, ST5 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
308944, 6764, 76954 | |
Antibody | |
IgG |
Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P78524, Q924W7 | |
St5 | |
ST5 Fusion Protein Ag3403 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |